Learn More
Tocris Bioscience™ ML 339
GREENER_CHOICE
Product Code. 18759738
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18759738
Brand: Tocris Bioscience™ 5943/10
Potent and selective hCXCR6 antagonist
ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide
CAS 2080300-49-6
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Target Chemokine Receptor Antagonists
Molecular Formula C26H32ClN3O5
Formula Weight 502
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity 98%
Show More Show Less
For Research Use Only.