missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitoferrin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Mitoferrin 1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884925
|
Proteintech
26469-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874925
|
Proteintech
26469-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.Specifications
Mitoferrin 1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q920G8, Q9NYZ2 | |
51312, 67712 | |
Mitoferrin 1 Fusion Protein Ag24876 | |
Primary | |
-20°C | |
SLC25A37 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MFRN, Mfrn1, Mitochondrial iron transporter 1, Mitoferrin 1, SLC25A37 | |
SLC25A37 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |