Learn More
Ansell MICROFLEX™ TQ-601 Soft Moisturizing Nitrile Gloves
Product Code. 17803692 Shop All Ansell Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
Product Code. 17803692
Brand: Ansell TQ601XL
White nitrile exam gloves designed with HYDRASOFT™ Skin Moisturizing Technology to keep away dryness during medical procedures.
  • ASTM D6319, FDA 510(k), CE Number certified
  • Gloves comfortably protect hands while replenishing moisture and preventing dryness
  • Patented HYDRASOFT™ Skin Moisturizing Technology is proven to allow skin to retain 2X more moisture* than gloves without HYDRASOFT™
  • Super soft, lightweight nitrile construction provides extra comfort and tactility while working
*Based on internal testing data on file.
Specifications
Certifications/Compliance ASTM D6319, CE Number, FDA21 CFR 177-2600-US Food Contact Approved, US QSR Medical Examination Grade
Certifications/Compliance (Europe) EN 455 1-4, EN ISO 13485
Accelerator Free No
Applications Healthcare, Laboratory
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4.7 mil
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.8 mil
Palm Thickness (Metric) 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (English) 2.8 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Model MICROFLEX-TQ-601
Length (English) 9.6 in.
Length (Metric) 24.5 cm