Learn More
Duran™ Micro Filter Candle Without Tube
Designed for use with Duran™ filtration apparatus.
Brand:  Duran™ 258572107
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15282059
  • 2710.00 NOK / Pack of 10
Estimated Shipment
10-07-2019
Add to basket
Description
Description
  • Micro filter candle
  • Available with porosity of 0 to 4
  • Ten per pack
Specifications
Specifications
Dia: 13mm
10Pack
25mm
Videos