Learn More
Uvex Micro Black/Orange PU/RU 65291 Boot - Width 10
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine the limits for safety shoes; range in protection classes S3 and S1 P
1780.00 NOK
Specifications
Color | Black, Orange |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane rubber (PU/RU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17221451
|
Uvex
6529135 |
35 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17231451
|
Uvex
6529136 |
36 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17241451
|
Uvex
6529137 |
37 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17251451
|
Uvex
6529138 |
38 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17261451
|
Uvex
6529139 |
39 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17271451
|
Uvex
6529140 |
40 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17281451
|
Uvex
6529141 |
41 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17291451
|
Uvex
6529142 |
42 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17201461
|
Uvex
6529143 |
43 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17211461
|
Uvex
6529144 |
44 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17221461
|
Uvex
6529145 |
45 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17231461
|
Uvex
6529146 |
46 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17241461
|
Uvex
6529147 |
47 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17251461
|
Uvex
6529148 |
48 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17261461
|
Uvex
6529149 |
49 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17271461
|
Uvex
6529150 |
50 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17281461
|
Uvex
6529151 |
51 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17291461
|
Uvex
6529152 |
52 |
1780.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 VIBRAM Shoe 6529.1 S3 HI HRO SRC
- Sporty, multifunctional safety boots
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-repellent high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Reflecting elements for more safety
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue, dust tongue
- Injection-molded polyurethane scuffcap to protect upper leather on toe cap and significantly extend resistance time
- Meets ESD requirements: resistance to ground Kleiner
- Lightweight two-layer insole with excellent shock absorption characteristics and outstanding slip resistance
- Multilayer sole system comprising PU midsole and highly slip resistant rubber outersole from VIBRAM
- Optimized ankle protection to ensure safety when working on ladders
- Rugged, self-cleaning sole ridges ensure max grip for demanding applications on uneven or dirty floors (SRC labeling)
- Withstands brief exposure to heat approx. 300°C
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6529.1
- Standard EN ISO 20345:2011 S3 HI HRO SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane rubber (PU/RU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S3 |