missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA Antibody (MICA/4442) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14208JF525
This item is not returnable.
View return policy
Description
MICA Monoclonal antibody specifically detects MICA in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Protein Array
Specifications
| MICA | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FLJ60820, MGC111087, PERB11.1 | |
| Recombinant fragment (around aa1-200) of human MICA (exact sequence is proprietary) (Uniprot: Q29983 ) | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 100507436 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| MICA/4442 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction