missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mezlocillin sodium, 97%

Catalog Number p-7077806
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
42057-22-7
C21H24N5NaO8S2
561.56
BXRKBOLIXGCTKD-SYNJJEHYSA-N
131673974
(2S,5R,6R)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]
This item is not returnable. View return policy
42057-22-7
C21H24N5NaO8S2
561.56
BXRKBOLIXGCTKD-SYNJJEHYSA-N
131673974
(2S,5R,6R)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Formula Weight 561.56
Percent Purity 97%
Chemical Name or Material Mezlocillin sodium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'