missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-3779137
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
516-05-2
C4H6O4
118.09
MFCD00002656
ZIYVHBGGAOATLY-UHFFFAOYSA-N
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
487
CHEBI:30860
2-methylpropanedioic acid
CC(C(O)=O)C(O)=O
This item is not returnable. View return policy
516-05-2
C4H6O4
118.09
MFCD00002656
ZIYVHBGGAOATLY-UHFFFAOYSA-N
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
487
CHEBI:30860
2-methylpropanedioic acid
CC(C(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 129.0°C to 136.0°C
Color White
Density 1.4500g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula HO2CCH(CH3)CO2H
Quantity 5 g
Beilstein 02, IV, 1932
Specific Gravity 1.45
Solubility Information Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid
Formula Weight 118.09
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Methylmalonic acid
Show More Show Less
compliance-icons
Product Identifier
  • Methylmalonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'