missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7051844
Click to view available options
Quantity:
0.25 mol
11 mol
This item is not returnable. View return policy
676-58-4
CH3ClMg
74.79
MFCD00000468
RQNMYNYHBQQZSP-UHFFFAOYSA-M
methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
12670
magnesium;carbanide;chloride
C[Mg]Cl
This item is not returnable. View return policy
676-58-4
CH3ClMg
74.79
MFCD00000468
RQNMYNYHBQQZSP-UHFFFAOYSA-M
methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
12670
magnesium;carbanide;chloride
C[Mg]Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Brown to Green
Physical Form Liquid
Quantity 0.25 mol
Linear Formula CH3MgCl
UN Number UN3399
Beilstein 3587199
Flash Point -17° (1°F)
Solubility Information Miscible with diethyl ether and tetrahydrofuran.
Formula Weight 74.81
Concentration or Composition (by Analyte or Components) 3M in THF
Odor Ether-like
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
Density 1.013g/mL
Chemical Name or Material Methylmagnesium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'