missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfide, 97%, Thermo Scientific Chemicals

Catalog Number p-7043825
452.42 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-13-2
C8H10S
138.23
MFCD00008560
VHILIAIEEYLJNA-UHFFFAOYSA-N
methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
69334
1-methyl-4-methylsulfanylbenzene
CSC1=CC=C(C)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-13-2
C8H10S
138.23
MFCD00008560
VHILIAIEEYLJNA-UHFFFAOYSA-N
methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
69334
1-methyl-4-methylsulfanylbenzene
CSC1=CC=C(C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.029
Boiling Point 81°C to 82°C (18 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.573
Quantity 5 g
UN Number UN3334
Beilstein 1905733
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material Methyl p-tolyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'