Learn More
Methyl isothiocyanate, polymer-supported, 1.5-1.9 mmol/g on polystyrene, Thermo Scientific™
Catalog Number 11388786
Click to view available options
Quantity:
1 g
5 g
10 g
556-61-6
C2H3NS
73.11
MFCD00004818,MFCD03457480
LGDSHSYDSCRFAB-UHFFFAOYSA-N
methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide
11167
CHEBI:78337
methylimino(sulfanylidene)methane
CN=C=S
556-61-6
C2H3NS
73.11
MFCD00004818,MFCD03457480
LGDSHSYDSCRFAB-UHFFFAOYSA-N
methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide
11167
CHEBI:78337
methylimino(sulfanylidene)methane
CN=C=S
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Sensitivity Moisture sensitive
Chemical Name or Material Methyl isothiocyanate, polymer-supported, 1.5 to 1.9mmol/g on polystyrene