missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-3-carboxylate, 99%, Thermo Scientific Chemicals

Catalog Number p-7046478
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
942-24-5
C10H9NO2
175.187
MFCD00189407
QXAUTQFAWKKNLM-UHFFFAOYSA-N
methyl indole-3-carboxylate, indole-3-carboxylic acid methyl ester, methyl 3-indolecarboxylate, 1h-indole-3-carboxylic acid methyl ester, 1h-indole-3-carboxylic acid, methyl ester, 3-carbomethoxyindole, 3-methoxycarbonylindole, methylindole-3-carboxylate, indole-3-carboxylic acid, methyl ester, methyl-3-indolcarboxylate
589098
CHEBI:65019
methyl 1H-indole-3-carboxylate
COC(=O)C1=CNC2=CC=CC=C21
This item is not returnable. View return policy
942-24-5
C10H9NO2
175.187
MFCD00189407
QXAUTQFAWKKNLM-UHFFFAOYSA-N
methyl indole-3-carboxylate, indole-3-carboxylic acid methyl ester, methyl 3-indolecarboxylate, 1h-indole-3-carboxylic acid methyl ester, 1h-indole-3-carboxylic acid, methyl ester, 3-carbomethoxyindole, 3-methoxycarbonylindole, methylindole-3-carboxylate, indole-3-carboxylic acid, methyl ester, methyl-3-indolcarboxylate
589098
CHEBI:65019
methyl 1H-indole-3-carboxylate
COC(=O)C1=CNC2=CC=CC=C21

Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

Solubility
Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 149°C to 152°C
Quantity 25 g
Beilstein 142023
Solubility Information Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Methyl indole-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'