missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals

Catalog Number p-7055980
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
32807-28-6
C5H7ClO3
150.56
MFCD00000938
HFLMYYLFSNEOOT-UHFFFAOYSA-N
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
36240
methyl 4-chloro-3-oxobutanoate
COC(=O)CC(=O)CCl
This item is not returnable. View return policy
32807-28-6
C5H7ClO3
150.56
MFCD00000938
HFLMYYLFSNEOOT-UHFFFAOYSA-N
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042
36240
methyl 4-chloro-3-oxobutanoate
COC(=O)CC(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.305 g/mL
Boiling Point 85°C (4 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.456
Quantity 25 g
UN Number UN2922
Beilstein 1759939
Formula Weight 150.56
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Methyl 4-chloroacetoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'