Learn More
Methyl 3-phenylpropionate, 98%, Thermo Scientific Chemicals
Catalog Number p-7044217
Click to view available options
Quantity:
50 g
250 g
103-25-3
C10H12O2
164.204
MFCD00017209
RPUSRLKKXPQSGP-UHFFFAOYSA-N
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
7643
methyl 3-phenylpropanoate
COC(=O)CCC1=CC=CC=C1
103-25-3
C10H12O2
164.204
MFCD00017209
RPUSRLKKXPQSGP-UHFFFAOYSA-N
methyl 3-phenylpropionate, methyl hydrocinnamate, benzenepropanoic acid, methyl ester, 3-phenylpropionic acid methyl ester, methyl benzenepropanoate, hydrocinnamic acid, methyl ester, methyl dihydrocinnamate, methyl beta-phenylpropionate, methyl b-phenylpropionate, unii-111lc1gi10
7643
methyl 3-phenylpropanoate
COC(=O)CCC1=CC=CC=C1
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-phenylpropionate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Phenylpropionic Acid Methyl Ester is an ester derivative of 3-phenylpropionic acid that is sometimes present in fragrance ingredients. It is also used as an application, aromatic intermediate.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.045
Boiling Point 236°C to 239°C
Flash Point 100°C (212°F)
Refractive Index 1.502
Quantity 50 g
Beilstein 1909591
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Methyl 3-phenylpropionate