Learn More
Methyl 3-hexenoate, 95+%, predominantly trans, Thermo Scientific™
Catalog Number p-3593457
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2396-78-3
C7H12O2
128.17
MFCD00672771
XEAIHUDTEINXFG-SNAWJCMRSA-N
3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
5362782
methyl (E)-hex-3-enoate
CCC=CCC(=O)OC
2396-78-3
C7H12O2
128.17
MFCD00672771
XEAIHUDTEINXFG-SNAWJCMRSA-N
3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
5362782
methyl (E)-hex-3-enoate
CCC=CCC(=O)OC
Color Colorless to Yellow
Density 0.9150g/mL
Boiling Point 169°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CHCHCH2COOCH3
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.915
Solubility Information Solubility in water: practically insoluble
Formula Weight 128.17
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material Methyl 3-hexenoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 3-hexenoate, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.