missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals
1845.69 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.
Solubility
Immiscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.
Solubility
Immiscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Density | 1.53 |
Boiling Point | 58°C to 60°C (12 mmHg) |
Flash Point | 75°C (167°F) |
Refractive Index | 1.458 |
Quantity | 100 g |
Beilstein | 1742949 |
Solubility Information | Immiscible in water. |
Formula Weight | 167.01 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-bromopropionate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'