missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dioxo-4-(2-thienyl)butyrate, 97%, Thermo Scientific™

Catalog Number 15447928
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
57409-51-5
C9H8O4S
212.22
MFCD01249731
TZNHHDJRXUBHRB-ALCCZGGFSA-N
methyl 2,4-dioxo-4-thiophen-2-yl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid methyl ester, methyl 2,4-dioxo-4-thien-2-ylbutanoate, methyl 2,4-dioxo-4-2-thienyl butanoate, methyl 2,4-dioxo-4-2-thienyl butyrate, methyl 2-thienoylpyruvate, methyl 4-2-thienyl-2,4-dioxobutanoate, methyl 2,4-dioxo-4-thien-2-yl butanoate, methyl2,4-dioxo-4-thiophen-2-yl butanoate, 2-thiophenebutanoicacid, a,g-dioxo-, methyl ester
2771673
methyl 2,4-dioxo-4-thiophen-2-ylbutanoate
COC(=O)C(\O)=C\C(=O)C1=CC=CS1
This item is not returnable. View return policy
57409-51-5
C9H8O4S
212.22
MFCD01249731
TZNHHDJRXUBHRB-ALCCZGGFSA-N
methyl 2,4-dioxo-4-thiophen-2-yl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid methyl ester, methyl 2,4-dioxo-4-thien-2-ylbutanoate, methyl 2,4-dioxo-4-2-thienyl butanoate, methyl 2,4-dioxo-4-2-thienyl butyrate, methyl 2-thienoylpyruvate, methyl 4-2-thienyl-2,4-dioxobutanoate, methyl 2,4-dioxo-4-thien-2-yl butanoate, methyl2,4-dioxo-4-thiophen-2-yl butanoate, 2-thiophenebutanoicacid, a,g-dioxo-, methyl ester
2771673
methyl 2,4-dioxo-4-thiophen-2-ylbutanoate
COC(=O)C(\O)=C\C(=O)C1=CC=CS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN1325
Formula Weight 212.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2,4-dioxo-4-(2-thienyl)butyrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'