Learn More
Phenothrin in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Specifications
Quantity | 100 mL |
Type | ICP/MS Standards |
Concentration or Composition (by Analyte or Components) | Barium (Ba) 1000mg/L in HCl 2% |
Solvent or Matrix | HCl |