missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disulfoton-sulfone in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 100μg/mL |
CAS | 2497-06-5 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C8H19O4PS3 |
InChI Key | BKVJOVPVLOJPKJ-UHFFFAOYSA-N |
SMILES | CCOP(=S)(OCC)SCCS(=O)(=O)CC |
IUPAC Name | diethoxy-(2-ethylsulfonylethylsulfanyl)-sulfanylidene-$l^{5}-phosphane |
Molecular Weight (g/mol) | 306.39 |
PubChem CID | 17241 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'