Learn More
Dieldrin in Methanol 10μg/mL, Fisher Chemical™
Catalog Number 12925464
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 10μg/mL
CAS 60-57-1
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C12H8Cl6O
InChI Key DFBKLUNHFCTMDC-YNXGVQRSSA-N
SMILES C1C2C3C(C1C4C2O4)C5(C(=C(C3(C5(Cl)Cl)Cl)Cl)Cl)Cl
Molecular Weight (g/mol) 380.895
PubChem CID 101820673
Solvent or Matrix Methanol
Chemical Name or Material Dieldrin