Learn More
Chlorobenzilate in Methanol 10μg/mL, Fisher Chemical™
Catalog Number 12952444
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 10μg/mL
CAS 510-15-6
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C16H14Cl2O3
InChI Key RAPBNVDSDCTNRC-UHFFFAOYSA-N
SMILES CCOC(=O)C(C1=CC=C(C=C1)Cl)(C2=CC=C(C=C2)Cl)O
IUPAC Name ethyl 2,2-bis(4-chlorophenyl)-2-hydroxyacetate
Molecular Weight (g/mol) 325.185
PubChem CID 10522
ChEBI CHEBI:34626
Solvent or Matrix Methanol
Chemical Name or Material Chlorobenzilate