Learn More
Aldicarb in Acetonitrile 100μg/mL, Fisher Chemical™
Catalog Number 12929493
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 100μg/mL
CAS 116-06-3
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C7H14N2O2S
UN Number 1648
InChI Key QGLZXHRNAYXIBU-WEVVVXLNSA-N
SMILES CC(C)(C=NOC(=O)NC)SC
IUPAC Name [(E)-(2-methyl-2-methylsulfanylpropylidene)amino] N-methylcarbamate
Molecular Weight (g/mol) 190.261
PubChem CID 9570071
Solvent or Matrix Acetonitrile
Physical Form Liquid
Chemical Name or Material Aldicarb
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Aldicarb,2-methyl-2-(methylthio)propanal-O-(N-methylcarbamoyl)oxime 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .