Learn More
meta-Phosphoric acid, 65% HPO3 basis, Honeywell Fluka™
Catalog Number p-7120568
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
37267-86-0
HO3P
79.979
MFCD00011351
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
3084658
OP(=O)=O
37267-86-0
HO3P
79.979
MFCD00011351
UEZVMMHDMIWARA-UHFFFAOYSA-N
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
3084658
OP(=O)=O
Purum, ∼65% HPO3 basis, pieces
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Colorless
Melting Point 200°c
Boiling Point 600°C
Density 2g/mL
Packaging Poly bottle
Quantity 100 g
Linear Formula (HPO3)n
Physical Form Solid
UN Number UN3260
Percent Purity 65% (HPO3 basis)
Chemical Name or Material meta-Phosphoric acid
Show More Show Less
compliance-icons
Product Identifier
  • meta-Phosphoric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.