Learn More
Mesalazine Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Mesalazine Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Mesalazine Ethyl Ester |
| CAS | 62773-65-3 |
| Analyte or Component Names | Mesalazine Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Mesalazine Ethyl Ester, 5-Amino-2-hydroxybenzoic acid ethyl ester, Ethyl 2-hydroxy-5-aminobenzoate, Ethyl 5-amino-2-hydroxybenzoate, Ethyl 5-aminosalicylate, Ethyl m-aminosalicylate |
| InChI Formula | InChI=1S/C9H11NO3/c1-2-13-9(12)7-5-6(10)3-4-8(7)11/h3-5,11H,2,10H2,1H3 |
| SMILES | CCOC(=O)c1cc(N)ccc1O |
| IUPAC Name | ethyl 5-amino-2-hydroxybenzoate |
| Show More |