missing translation for 'onlineSavingsMsg'
Learn More

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents

Catalog Number p-27041
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass
This item is not returnable. View return policy
156-57-0
C2H8ClNS
113.60
MFCD00012904
OGMADIBCHLQMIP-UHFFFAOYSA-N
cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
9082
2-aminoethanethiol;hydrochloride
[H+].[Cl-].NCCS
This item is not returnable. View return policy
156-57-0
C2H8ClNS
113.60
MFCD00012904
OGMADIBCHLQMIP-UHFFFAOYSA-N
cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
9082
2-aminoethanethiol;hydrochloride
[H+].[Cl-].NCCS
β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
TRUSTED_SUSTAINABILITY
Melting Point 66°C
Color White
pH 3.3 to 5.0
Assay Percent Range 98%
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2776
Formula Weight 113.61
Percent Purity ∽98%
Physical Form Solid
Chemical Name or Material β-Mercaptoethylamine Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethanethiol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'