Learn More
Mepazine Hydrochloride Monohydrate, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Mepazine Hydrochloride Monohydrate |
| CAS | 6146-88-9 |
| Analyte or Component Names | Mepazine Hydrochloride Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N2 S . Cl H . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine, 10-[(1-methyl-3-piperidinyl)methyl]-, hydrochloride, hydrate (1:1:1), 10H-Phenothiazine, 10-[(1-methyl-3-piperidinyl)methyl]-, monohydrochloride, monohydrate (9CI), Phenothiazine, 10-[(1-methyl-3-piperidyl)methyl]-, monohydrochloride, monohydrate (8CI), Mepazine hydrochloride monohydrate |
| InChI Formula | InChI=1S/C19H22N2S.ClH.H2O/c1-20-12-6-7-15(13-20)14-21-16-8-2-4-10-18(16)22-19-11-5-3-9-17(19)21;;/h2-5,8-11,15H,6-7,12-14H2,1H3;1H;1H2 |
| Solution Type | Neat |
| SMILES | O.Cl.CN1CCCC(CN2c3ccccc3Sc4ccccc24)C1 |
| Show More |