Learn More
Abnova™ MaxBead Protein A
MaxBead Protein A is magnetic bead designed for capturing of IgG or antigen via protein A-IgG-antigen interaction.
Brand:  Abnova™ U0084.5x1mL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16002388
  • 5212.23 NOK / Pack of 5
Estimated Shipment: 22-12-2021
Add to basket
Specifications
Purification
5 x 1 mL