missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS Antibody (58N3E1) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22378JF635
This item is not returnable.
View return policy
Description
MAVS Monoclonal antibody specifically detects MAVS in Human samples. It is validated for Western Blot
Specifications
| MAVS | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
| Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody. | |
| 0.1 mL | |
| Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase | |
| 57506 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot | |
| 58N3E1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction