missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK4 Recombinant Human Protein
MARK4 Recombinant Human Protein
Brand: Invitrogen PV3851
Additional Details : Weight : 0.05250kg
Description
MARK4 is a serine⁄threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells.Specifications
NP_113605 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
102.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MARK4 | |
MARK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μg/mL MAPKAP kinase substrate peptide (KKLNRTLSVA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
MARK4 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.36mg/mL | |
1385, 57787 | |
MARK4 Recombinant Human Protein | |
–80°C | |
1 Each | |
MARK4 Recombinant Human Protein | |
CAMKL | |
160nmoles of phosphate transferred to MAPKAP kinase substrate peptide (KKLNRTLSVA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
70% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.