Learn More
Marcasite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Alfa Aesar™
Important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Brand: Alfa Aesar 043363.06
Additional Details : CAS Number : 1309-36-0 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
1309-36-0 | |
119.965 | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
14788 | |
S=[Fe]=S |
FeS2 | |
MFCD00064690 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
bis(sulfanylidene)iron |
Specifications
Marcasite | |
FeS2 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
bis(sulfanylidene)iron | |
14788 | |
5g |
1309-36-0 | |
MFCD00064690 | |
Insoluble in water. | |
S=[Fe]=S | |
119.965 | |
Glass bottle |