missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K2 (MEKK2), GST Tag Recombinant Protein
Human MAP3K2 (MEKK2), GST Tag Recombinant Protein
16000.00 NOK - 69300.00 NOK
Specifications
Accession Number | AAF63496.3 |
---|---|
Molecular Weight (g/mol) | 97.2 kDa |
Gene Alias | MAP3K2 (MEKK2) |
KinaseFamily | STE11 Kinase Family |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448583
|
Invitrogen™
PR7408A |
100μg |
16000.00 NOK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10230533
|
Invitrogen™
PV4010 |
1mg |
69300.00 NOK
1mg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP3K2 (MEKK2) activates kinases involved in the MAP kinase signaling pathway.
Specifications
AAF63496.3 | |
MAP3K2 (MEKK2) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: STE Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
97.2 kDa | |
STE11 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |