missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANF Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76836JF635
This item is not returnable.
View return policy
Description
MANF Polyclonal antibody specifically detects MANF in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MANF | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 635 | |
| Arginine-Rich Protein, arginine-rich, mutated in early stage tumors, ARMET, ARP, mesencephalic astrocyte-derived neurotrophic factor, Protein ARMET | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the carboxy terminus of human MANF. The immunogen is located within the last 50 amino acids of MANF. Amino Acid Squence: KYAPKAASARTD | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, DNA Repair | |
| 7873 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction