Learn More
Malonyl dichloride, 97%, Thermo Scientific Chemicals
Catalog Number p-3212841
677.00 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1663-67-8
C3H2Cl2O2
140.95
MFCD00000735
SXYFKXOFMCIXQW-UHFFFAOYSA-N
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
74269
propanedioyl dichloride
C(C(=O)Cl)C(=O)Cl
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1663-67-8
C3H2Cl2O2
140.95
MFCD00000735
SXYFKXOFMCIXQW-UHFFFAOYSA-N
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
74269
propanedioyl dichloride
C(C(=O)Cl)C(=O)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Brown to Yellow
Density 1.4500g/mL
Boiling Point 47.0°C (15.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCOCH2COCl
Refractive Index 1.4620 to 1.4660
Quantity 25 g
Beilstein 02, I, 252
Fieser 12,291
Specific Gravity 1.45
Solubility Information Solubility in water: reacts violently. Other solubilities: soluble in ether
Formula Weight 140.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Malonyl dichloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Malonyl dichloride
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.