missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Magnesium hyposulfite finds application in the treatment of certain skin diseases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 170°C (in -3H2O) |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Linear Formula | MgS2O3·6H2O |
Merck Index | 14,5694 |
Solubility Information | Soluble in water. |
Formula Weight | 244.53 (136.44 Anhydrous) |
Percent Purity | 99% |
Density | 1.818 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'