missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1261°C |
Color | White |
Physical Form | Powder |
Quantity | 2 g |
Linear Formula | MgF2 |
Merck Index | 14,5665 |
Solubility Information | Soluble in water,nitric acid. Insoluble in ethanol. |
Formula Weight | 62.3 |
Percent Purity | 99.99% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'