Learn More
Magnesium fluoride hydrate, min 87% MgF2, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.
Solubility
Soluble in water (87 mg/L at 18°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents and strong acids. Store at room temperature.
Magnesium fluoride is used in ceramics, optical lenses, glass manufacturing, and single crystals for polarizing prisms. It is also used in organic synthesis.
Solubility
Soluble in water (87 mg/L at 18°C ).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents and strong acids. Store at room temperature.
Specifications
Quantity | 500 g |
Merck Index | 14,5665 |
Solubility Information | Soluble in water (87mg/L at 18°C ). |
Formula Weight | 62.30 (Anhydrous) |
Odor | Odorless |
Assay | ≥87% MgF2 |
Chemical Name or Material | Magnesium fluoride hydrate |