missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18941-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Specifications
LSM7 | |
Polyclonal | |
Unconjugated | |
LSM7 | |
LSM7, YNL147W | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362829, 51690, 66094 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CQQ8, Q9UK45 | |
LSM7 | |
LSM7 Fusion Protein Ag13498 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |