missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM14B Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18490JF635
This item is not returnable.
View return policy
Description
LSM14B Polyclonal antibody specifically detects LSM14B in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| LSM14B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 149986 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 635 | |
| bA11M20.3, C20orf40, FAM61B, FT005, LSM13, LSM14B LSM14B, SCD6 homolog B (S. cerevisiae), RAP55B | |
| Recognizes region between residues Between between 335 and 385.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction