missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17750-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein related to the yeast large subunit GTPase 1. The encoded protein is necessary for cell viability and may localize in the endoplasmic reticulum, nucleus and cytoplasm.Specifications
LSG1 | |
Polyclonal | |
Unconjugated | |
LSG1 | |
hLsg1, Large subunit GTPase 1 homolog, LSG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
224092, 288029, 55341 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UM18, Q5BJT6, Q9H089 | |
Lsg1 | |
LSG1 Fusion Protein Ag12016 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |