Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Manganese
For use with MC500 multi-parameter colorimeter
Brand: Lovibond™ 517621BT
Additional Details : Weight : 0.30000kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Includes:
- Manganese LR No. 1 Tablets, 100 pack
- Manganese LR No. 2 Tablets, 100 pack
Description
- Parameter: Manganese
- Method: Formaldoxime
- Sample Size: 10mL
Specifications
Tablet | |
MC500 multi-parameter colorimeter | |
100 | |
Formaldoxime |
Water quality testing | |
Manganese No. 1 LR tablets (100 pack), manganese No. 2 LR tablets (100 pack) | |
0.02 to 4mg/L | |
Manganese Reagent Set |