Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine
For use with MC500 multi-parameter colorimeter
Brand: Lovibond™ 471056
Additional Details : Weight : 0.10000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
For use with MC500 multi-parameter colorimeter
- Parameter: Chlorine
- Method: DPD
Specifica
Water quality testing | |
15mL DPD No. 1 buffer solution, 15mL DPD No. 1 reagent solution, 15mL DPD No. 3 reagent solution | |
0.02 to 4mg/L | |
Chlorine Reagent Set |
MC500 multi-parameter colorimeter | |
50 | |
DPD |