Learn More
Lonza™ FlashGel™ System Kit
Fast analysis, recovery, and documentation of DNA and RNA
Brand: Lonza™ 57067
Additional Details : Weight : 6.80390kg
Alert:
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Description
- Provides nearly instant results
- Load samples, watch bands migrate and get data in as little as two minutes
- Includes DNA recovery and gel documentation
- Needs no gel preparation, band excision, purification, and UV light
- Complete separation, recovery and documentation safely at the bench
- Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
- Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
- Detect <0.1ng DNA or <10ng total RNA
- Clean, sharp separation and straight, uniform sample lanes
FlashGel System
- Consists of enclosed, disposable, precast agarose gel cassettes and a combination unit
- Includes: FlashGel Dock; FlashGel Camera; 9 Pk. FlashGel DNA Cassettes (1.2%, 12 + 1 well); FlashGel Loading Dye and FlashGel DNA Marker
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
FlashGel System | |
FlashGel Dock, a FlashGel Camera, a 9 Pk. of 1.2% DNA Cassettes, Loading Dye and Marker |