Learn More
Lonza™ BioWhittaker™ BME with Earle's BSS, without L-glutamine
Historical precursor to Minimum Essential Medium (MEM)
Brand: Lonza™ BE12-105F
Additional Details : Weight : 0.64000kg
Description
- Suitable for variety of cell types
- Manufactured in accordance with cGMP regulations
- Formulations recommended by the Tissue Culture Association
Specifications
15° to 30°C | |
Basal Medium | |
No Glutamine | |
500 mL |