Learn More
Lonza™ BioWhittaker™ Alpha MEM Eagle Medium
Used to culture bone marrow cells and amniotic cells for chromosome analysis. BioWhittaker™ Alpha MEM Eagle Medium are manufactured in accordance with cGMP regulations.
Brand: Lonza™
Additional Details : Weight : 0.64000kg
Description
Differs from standard MEM by containing the following unique components:
- Vitamin B12
- Ascorbic acid
- Non-essential amino acids
- Sodium pyruvate
- Lipoic acid
- D-Biotin
Specifications
2° to 8°C | |
Sterile-filtered through two consecutive 0.2 μm filters | |
6.91 to 7.27 |
Sterile | |
No Glutamine, No Deoxyribonucleosides, No Ribonucleosides | |
500 mL |