Learn More
Loading Buffer, Dual Color, Prosieve™ ProTrack™, Lonza
Brand:  Lonza 193861
Additional Details : Weight : 0.45000kg
Product Code. 11528005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Loading Buffer
Prosieve ProTrack Dual Color
1000 Samples
Consists of 1 vial of 5mL (4x Dual Color Protein Loading Buffer) and 1 vial of 1.0mL (20X Reducing Agent (2M DTT)), sufficient for 1000 samples of 20μL (or equivalent)