Learn More
Loading Buffer, Dual Color, Prosieve™ ProTrack™, Lonza
Brand: Lonza 193861
Additional Details : Weight : 0.45000kg
Specifications
Loading Buffer | |
Prosieve ProTrack Dual Color | |
1000 Samples | |
Consists of 1 vial of 5mL (4x Dual Color Protein Loading Buffer) and 1 vial of 1.0mL (20X Reducing Agent (2M DTT)), sufficient for 1000 samples of 20μL (or equivalent) |