missing translation for 'onlineSavingsMsg'
Learn More

Lithium metaborate, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number p-222312
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
lithium;oxido(oxo)borane
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
lithium;oxido(oxo)borane
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 845°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula LiBO2
Solubility Information Solubility in water: 25.7g/L (20°C)
Formula Weight 49.75
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Lithium metaborate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'