missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Bromide Anhydrous, Extra Pure, SLR, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Spezifikation
Boiling Point | 1265°C |
Melting Point | 550°C |
Color | Beige |
pH | 7 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Formula Weight | 86.84g/mol |
Packaging | Plastic powder jar |
Sicherheit und Handhabung
Produktidentifikator
- Lithium bromide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'