Learn More
Lithium aluminum di-n-butylamide, 0.16M solution in 1,2-Dimethoxyethane, Thermo Scientific Chemicals
Catalog Number p-4813513
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
15405-86-4
C32H72AlLiN4
546.87
ISBVOXPIPNIBOM-UHFFFAOYSA-N
lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
57369578
lithium;tetrakis(dibutylamino)alumanuide
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC
15405-86-4
C32H72AlLiN4
546.87
ISBVOXPIPNIBOM-UHFFFAOYSA-N
lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
57369578
lithium;tetrakis(dibutylamino)alumanuide
[Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -54°C
Quantity 100 mL
Flash Point -6°C
Formula Weight 546.87
Packaging Glass bottle
Specific Gravity 0.86
Density 0.8600g/mL
Chemical Name or Material Lithium Aluminum di-n-butylamide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium aluminium di-n-butylamide, 0.16M solution in 1,2-dimethoxyethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.