missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Listeria Monocytogenes Antibody (LZF7) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB600-1281JF525
3871.75 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Listeria Monocytogenes Monoclonal antibody specifically detects Listeria Monocytogenes in Bacteria samples. It is validated for Western Blot,Sandwich ELISA
Specifications
| Listeria Monocytogenes | |
| Monoclonal | |
| Janelia Fluor 525 | |
| L. monocytogenes | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified L. monocytogenes outer membrane fraction (OM). | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Sandwich ELISA | |
| LZF7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction