missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN28 Mouse anti-Human, Clone: 4F5G6, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | LIN28 |
---|---|
Clone | 4F5G6 |
Concentration | 1.56 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887455
|
Proteintech
60344-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877455
|
Proteintech
60344-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.Specifications
LIN28 | |
1.56 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CSDD1, LIN 28, lin 28 homolog (C. elegans), Lin 28A, LIN28, Lin28A, Protein lin 28 homolog A, ZCCHC1 | |
LIN28A | |
IgG1 | |
Protein G | |
Antibody |
4F5G6 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q9H9Z2 | |
79727 | |
LIN28 Fusion Protein Ag2312 | |
Primary | |
-20°C | |
LIN28A |